The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006833.4 | NP_006824.2 | 1 - 2 | 110 | 59 |
GenBank mRNA | AK295237.1 | - | 1 - 2 | 88 | 59 |
AK312024.1 | - | 1 - 2 | 101 | 59 | |
BC002520.2 | - | 1 - 2 | 84 | 59 | |
BI113801.1 | - | 1 - 2 | 110 | 59 | |
CB990467.1 | - | 1 - 2 | 134 | 59 |