The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080512.2 | NP_001073981.1 | 3 - 4 | 302 | 108 |
XM_005270169.4 | XP_005270226.1 | 2 - 3 | 3300 | 108 | |
XM_011540185.2 | XP_011538487.1 | 4 - 5 | 548 | 108 | |
XM_011540188.2 | XP_011538490.2 | 3 - 4 | 433 | 108 | |
XM_011540189.2 | XP_011538491.1 | 2 - 3 | 199 | 108 | |
XM_011540190.2 | XP_011538492.1 | 2 - 3 | 156 | 108 | |
XM_017016677.1 | XP_016872166.1 | 4 - 5 | 6936 | 108 | |
XM_017016678.1 | XP_016872167.1 | 5 - 6 | 453 | 108 | |
GenBank mRNA | AY726586.1 | - | 3 - 4 | 247 | 108 |