The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080512.2 | NP_001073981.1 | 19 - 20 | 2693 | 67 |
XM_005270169.4 | XP_005270226.1 | 18 - 19 | 5691 | 67 | |
XM_011540185.2 | XP_011538487.1 | 20 - 21 | 2939 | 67 | |
XM_011540188.2 | XP_011538490.2 | 19 - 20 | 2824 | 67 | |
XM_011540189.2 | XP_011538491.1 | 18 - 19 | 2590 | 67 | |
XM_011540190.2 | XP_011538492.1 | 18 - 19 | 2547 | 67 | |
XM_011540191.2 | XP_011538493.1 | 16 - 17 | 2409 | 67 | |
XM_017016677.1 | XP_016872166.1 | 20 - 21 | 9327 | 67 | |
XM_017016678.1 | XP_016872167.1 | 21 - 22 | 2844 | 67 | |
GenBank mRNA | AY726586.1 | - | 19 - 20 | 2639 | 67 |
BM972104.1 | - | 2 - 3 | 67 |