Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR3
Entrez Gene ID
55027
Gene Name
HEAT repeat containing 3
Gene Aliases
SYO1
Chromosome Location
Chr.16: 50065970 - 50107268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182922.2 NP_891552.1 2 - 3 480 83
XM_017023385.1 XP_016878874.1 1 - 2 543 83
GenBank mRNA AK289949.1 - 2 - 3 445 83
BC014415.1 - - 457 83
BC018730.1 - 2 - 3 445 83
BC033077.1 - 2 - 3 346 83

View Full Product Details