The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100399.1 | NP_001093869.1 | 30 - 31 | 4083 | 97 |
XM_011513672.2 | XP_011511974.1 | 30 - 31 | 3696 | 97 | |
XM_017007928.1 | XP_016863417.1 | 30 - 31 | 3658 | 97 | |
GenBank mRNA | AB014548.1 | - | 18 - 19 | 2087 | 97 |
AB621803.1 | - | 30 - 31 | 4097 | 97 | |
AF294791.1 | - | 30 - 31 | 3654 | 97 | |
AK021757.1 | - | 5 - 6 | 569 | 97 | |
AK023592.1 | - | 3 - 4 | 336 | 97 | |
AK124569.1 | - | 2 - 3 | 2885 | 97 | |
BC009650.1 | - | 6 - 7 | 668 | 97 | |
BC114218.1 | - | 30 - 31 | 3779 | 97 | |
BC126225.1 | - | 29 - 30 | 3493 | 97 | |
CR749425.1 | - | 29 - 30 | 3452 | 97 |