Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAF8
Entrez Gene ID
22828
Gene Name
SR-related CTD associated factor 8
Gene Aliases
RBM16
Chromosome Location
Chr.6: 154733378 - 154834244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286188.1 NP_001273117.1 17 - 18 2205 89
NM_001286189.1 NP_001273118.1 18 - 19 2169 89
NM_001286194.1 NP_001273123.1 17 - 18 2106 89
NM_001286199.1 NP_001273128.1 17 - 18 2158 89
NM_014892.4 NP_055707.3 16 - 17 2449 89
GenBank mRNA AB029039.1 - 16 - 17 2111 89
AK295651.1 - 17 - 18 2167 89
AK302969.1 - 17 - 18 2091 89
AK303001.1 - 18 - 19 2131 89
AK307815.1 - 18 - 19 2139 89
BC070071.1 - 17 - 18 2158 89

View Full Product Details