The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286188.1 | NP_001273117.1 | 6 - 7 | 751 | 68 |
NM_001286189.1 | NP_001273118.1 | 7 - 8 | 715 | 68 | |
NM_001286194.1 | NP_001273123.1 | 6 - 7 | 652 | 68 | |
NM_001286199.1 | NP_001273128.1 | 6 - 7 | 704 | 68 | |
NM_014892.4 | NP_055707.3 | 5 - 6 | 995 | 68 | |
GenBank mRNA | AB029039.1 | - | 5 - 6 | 657 | 68 |
AK295651.1 | - | 6 - 7 | 713 | 68 | |
AK302969.1 | - | 6 - 7 | 637 | 68 | |
AK303001.1 | - | 7 - 8 | 677 | 68 | |
AK307815.1 | - | 7 - 8 | 685 | 68 | |
BC032728.1 | - | 5 - 6 | 957 | 68 | |
BC070071.1 | - | 6 - 7 | 704 | 68 |