The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286609.1 | NP_001273538.1 | 9 - 10 | 1456 | 97 |
NM_032131.5 | NP_115507.4 | 10 - 11 | 1529 | 97 | |
XM_005267154.3 | XP_005267211.1 | 10 - 11 | 1810 | 97 | |
XM_005267155.2 | XP_005267212.1 | 10 - 11 | 1545 | 97 | |
XM_005267157.3 | XP_005267214.1 | 10 - 11 | 1453 | 97 | |
XM_006715573.2 | XP_006715636.1 | 11 - 12 | 1635 | 97 | |
XM_006715574.2 | XP_006715637.1 | 7 - 8 | 965 | 97 | |
XM_011536166.1 | XP_011534468.1 | 11 - 12 | 1620 | 97 | |
XM_011536167.1 | XP_011534469.1 | 10 - 11 | 1523 | 97 | |
XM_011536168.2 | XP_011534470.1 | 10 - 11 | 1523 | 97 | |
XM_011536170.2 | XP_011534472.1 | 10 - 11 | 1523 | 97 | |
XM_011536172.2 | XP_011534474.1 | 9 - 10 | 1380 | 97 | |
XM_011536176.2 | XP_011534478.1 | 10 - 11 | 1523 | 97 | |
XM_017011350.1 | XP_016866839.1 | 9 - 10 | 1458 | 97 | |
XM_017011351.1 | XP_016866840.1 | 10 - 11 | 1523 | 97 | |
XM_017011352.1 | XP_016866841.1 | 9 - 10 | 1359 | 97 | |
GenBank mRNA | AK095234.1 | - | 9 - 10 | 1372 | 97 |
AK292499.1 | - | 10 - 11 | 1523 | 97 | |
AK294574.1 | - | 9 - 10 | 1456 | 97 | |
AL136754.1 | - | 6 - 7 | 879 | 97 | |
BC030603.2 | - | 10 - 11 | 1523 | 97 |