The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286609.1 | NP_001273538.1 | 5 - 6 | 852 | 60 |
NM_032131.5 | NP_115507.4 | 6 - 7 | 925 | 60 | |
XM_005267154.3 | XP_005267211.1 | 6 - 7 | 1206 | 60 | |
XM_005267155.2 | XP_005267212.1 | 6 - 7 | 941 | 60 | |
XM_005267157.3 | XP_005267214.1 | 6 - 7 | 849 | 60 | |
XM_006715573.2 | XP_006715636.1 | 7 - 8 | 1031 | 60 | |
XM_006715574.2 | XP_006715637.1 | 3 - 4 | 361 | 60 | |
XM_011536166.1 | XP_011534468.1 | 7 - 8 | 1016 | 60 | |
XM_011536167.1 | XP_011534469.1 | 6 - 7 | 919 | 60 | |
XM_011536168.2 | XP_011534470.1 | 6 - 7 | 919 | 60 | |
XM_011536170.2 | XP_011534472.1 | 6 - 7 | 919 | 60 | |
XM_011536172.2 | XP_011534474.1 | 5 - 6 | 776 | 60 | |
XM_011536176.2 | XP_011534478.1 | 6 - 7 | 919 | 60 | |
XM_017011351.1 | XP_016866840.1 | 6 - 7 | 919 | 60 | |
XM_017011352.1 | XP_016866841.1 | 6 - 7 | 931 | 60 | |
GenBank mRNA | AK095234.1 | - | 5 - 6 | 768 | 60 |
AK292499.1 | - | 6 - 7 | 919 | 60 | |
AK294574.1 | - | 5 - 6 | 852 | 60 | |
AL136754.1 | - | 2 - 3 | 275 | 60 | |
BC030603.2 | - | 6 - 7 | 919 | 60 |