The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324208.1 | NP_001311137.1 | 6 - 7 | 935 | 149 |
NM_001324213.1 | NP_001311142.1 | 6 - 7 | 1115 | 149 | |
NM_021952.4 | NP_068771.2 | 6 - 7 | 1045 | 149 | |
XM_006710411.3 | XP_006710474.1 | 6 - 7 | 1027 | 149 | |
XM_011540889.2 | XP_011539191.1 | 6 - 7 | 910 | 149 | |
XM_011540893.2 | XP_011539195.1 | 6 - 7 | 857 | 149 | |
XM_011540894.2 | XP_011539196.1 | 7 - 8 | 1199 | 149 | |
XM_011540895.2 | XP_011539197.1 | 7 - 8 | 1103 | 149 | |
GenBank mRNA | AY033998.1 | - | 6 - 7 | 999 | 149 |
M62843.1 | - | 6 - 7 | 869 | 149 |