The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022370.3 | NP_071765.2 | 19 - 20 | 3017 | 92 |
XM_017018122.1 | XP_016873611.1 | 18 - 19 | 2782 | 92 | |
GenBank mRNA | AK024697.1 | - | 3 - 4 | 160 | 92 |
AK225076.1 | - | 3 - 4 | 160 | 92 | |
AK291045.1 | - | 19 - 20 | 2951 | 92 | |
AK295475.1 | - | 3 - 4 | 156 | 92 | |
AY509035.1 | - | 19 - 20 | 3040 | 92 |