Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FEZ1
Entrez Gene ID
9638
Gene Name
fasciculation and elongation protein zeta 1
Gene Aliases
UNC-76
Chromosome Location
Chr.11: 125445745 - 125496310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005103.4 NP_005094.1 6 - 7 1171 74
XM_005271734.2 XP_005271791.1 6 - 7 1216 74
XM_005271735.2 XP_005271792.1 6 - 7 1268 74
GenBank mRNA BC009545.1 - 6 - 7 1088 74
CR456817.1 - 5 - 6 936 74
CR541954.1 - 5 - 6 936 74
U60060.1 - 6 - 7 1035 74
U69139.1 - 5 - 6 961 74

View Full Product Details