Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FEZ1
Entrez Gene ID
9638
Gene Name
fasciculation and elongation protein zeta 1
Gene Aliases
UNC-76
Chromosome Location
Chr.11: 125445745 - 125496310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005103.4 NP_005094.1 8 - 9 1330 76
XM_005271734.2 XP_005271791.1 8 - 9 1375 76
XM_005271735.2 XP_005271792.1 8 - 9 1427 76
GenBank mRNA BC009545.1 - 8 - 9 1247 76
CR456817.1 - 7 - 8 1095 76
CR541954.1 - 7 - 8 1095 76
U60060.1 - 8 - 9 1194 76
U69139.1 - 7 - 8 1120 76

View Full Product Details