Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
Chromosome Location
Chr.11: 123072006 - 123195305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024769.3 NP_079045.1 5 - 6 1048 62
GenBank mRNA AK026068.1 - 5 - 6 1024 62
AK304843.1 - 5 - 6 768 62
AY326422.1 - 5 - 6 872 62
AY358340.1 - 5 - 6 872 62
BC009371.2 - 5 - 6 694 62

View Full Product Details