Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIXDC1
Entrez Gene ID
85458
Gene Name
DIX domain containing 1
Gene Aliases
CCD1
Chromosome Location
Chr.11: 111927144 - 112022650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037954.3 NP_001033043.1 7 - 8 1216 119
NM_033425.4 NP_219493.1 3 - 4 749 119
XM_017018466.1 XP_016873955.1 8 - 9 1176 119
XM_017018467.1 XP_016873956.1 8 - 9 1044 119
XM_017018468.1 XP_016873957.1 3 - 4 360 119
GenBank mRNA AB051522.1 - 3 - 4 656 119
AK055899.1 - 1 - 2 272 119
AK289807.1 - 3 - 4 725 119
BC033034.2 - 3 - 4 708 119
DQ642016.1 - 7 - 8 1216 119

View Full Product Details