Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIXDC1
Entrez Gene ID
85458
Gene Name
DIX domain containing 1
Gene Aliases
CCD1
Chromosome Location
Chr.11: 111927144 - 112022650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037954.3 NP_001033043.1 13 - 14 1667 82
NM_033425.4 NP_219493.1 9 - 10 1200 82
XM_017018466.1 XP_016873955.1 14 - 15 1627 82
XM_017018467.1 XP_016873956.1 14 - 15 1495 82
XM_017018468.1 XP_016873957.1 9 - 10 811 82
XM_017018469.1 XP_016873958.1 10 - 11 836 82
GenBank mRNA AB051522.1 - 9 - 10 1107 82
AK055899.1 - 7 - 8 723 82
AK289807.1 - 9 - 10 1176 82
AK294954.1 - 4 - 5 366 82
BC033034.2 - 9 - 10 1159 82
BC041626.1 - 6 - 7 394 82
DQ642016.1 - 13 - 14 1667 82

View Full Product Details