The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037954.3 | NP_001033043.1 | 9 - 10 | 1358 | 79 |
NM_033425.4 | NP_219493.1 | 5 - 6 | 891 | 79 | |
XM_017018466.1 | XP_016873955.1 | 10 - 11 | 1318 | 79 | |
XM_017018467.1 | XP_016873956.1 | 10 - 11 | 1186 | 79 | |
XM_017018468.1 | XP_016873957.1 | 5 - 6 | 502 | 79 | |
XM_017018469.1 | XP_016873958.1 | 6 - 7 | 527 | 79 | |
GenBank mRNA | AB051522.1 | - | 5 - 6 | 798 | 79 |
AK055899.1 | - | 3 - 4 | 414 | 79 | |
AK289807.1 | - | 5 - 6 | 867 | 79 | |
BC033034.2 | - | 5 - 6 | 850 | 79 | |
BC041626.1 | - | 2 - 3 | 85 | 79 | |
DQ642016.1 | - | 9 - 10 | 1358 | 79 |