The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025003.3 | NP_079279.3 | 29 - 30 | 4453 | 78 |
XM_011538754.2 | XP_011537056.1 | 29 - 30 | 4506 | 78 | |
XM_017019979.1 | XP_016875468.1 | 22 - 23 | 3522 | 78 | |
GenBank mRNA | AF488804.1 | - | 29 - 30 | 4456 | 78 |
AJ515153.3 | - | 29 - 30 | 4447 | 78 |