Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFAT5
Entrez Gene ID
10725
Gene Name
nuclear factor of activated T-cells 5, tonicity-responsive
Gene Aliases
NF-AT5, NFATL1, NFATZ, OREBP, TONEBP
Chromosome Location
Chr.16: 69565952 - 69704666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138714.3 NP_619728.2 4 - 5 647 95
NM_173214.2 NP_775321.1 3 - 4 593 95
NM_173215.2 NP_775322.1 3 - 4 593 95
XM_011522820.2 XP_011521122.1 2 - 3 151 95
XM_017022871.1 XP_016878360.1 3 - 4 607 95
GenBank mRNA AF089824.1 - 3 - 4 535 95
AF346509.1 - 4 - 5 1533 95
AJ243298.1 - 3 - 4 352 95

View Full Product Details