The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_119376.1 | 26 - 27 | 3387 | 146 | |
GenBank mRNA | AY461812.1 | - | 26 - 27 | 3462 | 146 |