The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002748.3 | NP_002739.1 | 1 - 2 | 161 | 61 |
GenBank mRNA | BC035492.2 | - | 1 - 2 | 121 | 61 |
CR749401.1 | - | 1 - 2 | 138 | 61 | |
DA109756.1 | - | 1 - 2 | 161 | 61 | |
L77964.1 | - | 1 - 2 | 193 | 61 |