The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306177.1 | NP_001293106.1 | 17 - 18 | 1946 | 87 |
NM_004840.2 | NP_004831.1 | 18 - 19 | 2408 | 87 | |
XM_005262499.2 | XP_005262556.1 | 16 - 17 | 2573 | 87 | |
XM_011531412.2 | XP_011529714.1 | 19 - 20 | 2801 | 87 | |
XM_011531413.1 | XP_011529715.1 | 18 - 19 | 2732 | 87 | |
XM_011531414.1 | XP_011529716.1 | 17 - 18 | 2711 | 87 | |
XM_011531415.2 | XP_011529717.1 | 18 - 19 | 2156 | 87 | |
XM_011531416.2 | XP_011529718.1 | 18 - 19 | 2419 | 87 | |
XM_011531417.1 | XP_011529719.1 | 17 - 18 | 2350 | 87 | |
XM_017029975.1 | XP_016885464.1 | 19 - 20 | 2813 | 87 | |
XM_017029976.1 | XP_016885465.1 | 15 - 16 | 2191 | 87 | |
GenBank mRNA | AK291742.1 | - | 18 - 19 | 1951 | 87 |
AK294929.1 | - | 17 - 18 | 1752 | 87 | |
AK295709.1 | - | 18 - 19 | 2028 | 87 | |
BC033768.1 | - | 6 - 7 | 579 | 87 | |
BC039856.1 | - | 18 - 19 | 2408 | 87 | |
BC043505.1 | - | 17 - 18 | 1912 | 87 | |
BX537390.1 | - | 18 - 19 | 2013 | 87 | |
D13631.1 | - | 10 - 11 | 986 | 87 | |
D25304.1 | - | 18 - 19 | 1939 | 87 |