The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258333.1 | NP_001245262.1 | 10 - 11 | 1207 | 87 |
NM_001258334.1 | NP_001245263.1 | 11 - 12 | 1274 | 87 | |
NM_002108.3 | NP_002099.1 | 11 - 12 | 1274 | 87 | |
XM_011538249.2 | XP_011536551.1 | 2 - 3 | 107 | 87 | |
XM_017019246.1 | XP_016874735.1 | 2 - 3 | 595 | 87 | |
GenBank mRNA | AK298736.1 | - | 11 - 12 | 1253 | 87 |
AK303544.1 | - | 10 - 11 | 1133 | 87 | |
BC096096.1 | - | 10 - 11 | 1067 | 87 | |
BC096097.3 | - | 11 - 12 | 1134 | 87 | |
BC096098.3 | - | 11 - 12 | 1134 | 87 | |
BC096099.1 | - | 11 - 12 | 1134 | 87 | |
D16626.1 | - | 11 - 12 | 1148 | 87 |