The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018011.1 | NP_001018011.1 | 2 - 3 | 1381 | 68 |
NM_006006.4 | NP_005997.2 | 2 - 3 | 1536 | 68 | |
XM_005271658.4 | XP_005271715.1 | 2 - 3 | 1668 | 68 | |
XM_017018257.1 | XP_016873746.1 | 2 - 3 | 1469 | 68 | |
XM_017018258.1 | XP_016873747.1 | 2 - 3 | 1624 | 68 | |
XM_017018259.1 | XP_016873748.1 | 2 - 3 | 1976 | 68 | |
GenBank mRNA | AB208916.1 | - | 2 - 3 | 1520 | 68 |
BC026902.1 | - | 2 - 3 | 1440 | 68 | |
BC029812.1 | - | 2 - 3 | 1518 | 68 | |
BX648973.1 | - | 2 - 3 | 1653 | 68 | |
KU178400.1 | - | 1 - 2 | 1272 | 68 | |
KU178401.1 | - | 1 - 2 | 1272 | 68 | |
Z19002.1 | - | 1 - 2 | 1347 | 68 |