The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018011.1 | NP_001018011.1 | 5 - 6 | 1735 | 70 |
NM_006006.4 | NP_005997.2 | 5 - 6 | 1890 | 70 | |
XM_017018257.1 | XP_016873746.1 | 5 - 6 | 1823 | 70 | |
XM_017018258.1 | XP_016873747.1 | 5 - 6 | 1978 | 70 | |
XM_017018259.1 | XP_016873748.1 | 5 - 6 | 2330 | 70 | |
GenBank mRNA | BC026902.1 | - | 5 - 6 | 1794 | 70 |
BC029812.1 | - | 5 - 6 | 1872 | 70 | |
BX648973.1 | - | 5 - 6 | 2007 | 70 | |
KU178400.1 | - | 4 - 5 | 1626 | 70 | |
KU178401.1 | - | 3 - 4 | 1539 | 70 | |
Z19002.1 | - | 4 - 5 | 1701 | 70 |