Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB16
Entrez Gene ID
7704
Gene Name
zinc finger and BTB domain containing 16
Gene Aliases
PLZF, ZNF145
Chromosome Location
Chr.11: 114059576 - 114256765 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018011.1 NP_001018011.1 6 - 7 1899 65
NM_006006.4 NP_005997.2 6 - 7 2054 65
XM_017018257.1 XP_016873746.1 6 - 7 1987 65
XM_017018258.1 XP_016873747.1 6 - 7 2142 65
XM_017018259.1 XP_016873748.1 6 - 7 2494 65
GenBank mRNA BC026902.1 - 6 - 7 1958 65
BC029812.1 - 6 - 7 2036 65
BX648973.1 - 6 - 7 2171 65
KU178400.1 - 5 - 6 1790 65
KU178401.1 - 4 - 5 1703 65
Z19002.1 - 5 - 6 1865 65

View Full Product Details