Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCEA3
Entrez Gene ID
6920
Gene Name
transcription elongation factor A3
Gene Aliases
TFIIS, TFIIS.H
Chromosome Location
Chr.1: 23379953 - 23424779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003196.2 NP_003187.1 6 - 7 755 85
XM_006710864.1 XP_006710927.1 6 - 7 728 85
XM_011542053.2 XP_011540355.1 6 - 7 728 85
XM_017002200.1 XP_016857689.1 6 - 7 728 85
XM_017002201.1 XP_016857690.1 5 - 6 689 85
XM_017002202.1 XP_016857691.1 5 - 6 686 85
XM_017002203.1 XP_016857692.1 6 - 7 815 85
XM_017002204.1 XP_016857693.1 5 - 6 686 85
GenBank mRNA AJ223473.1 - 5 - 6 525 85
AK027024.1 - 6 - 7 716 85
AK290272.1 - 6 - 7 752 85
AK300707.1 - 6 - 7 705 85
AK310039.1 - 5 - 6 689 85
BC041613.1 - 6 - 7 638 85

View Full Product Details