The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308019.1 | NP_001294948.1 | 2 - 3 | 104 | 62 |
NM_015065.2 | NP_055880.2 | 1 - 2 | 231 | 62 | |
XM_017017397.1 | XP_016872886.1 | 2 - 3 | 120 | 62 | |
XM_017017398.1 | XP_016872887.1 | 2 - 3 | 120 | 62 | |
GenBank mRNA | AB014524.1 | - | 2 - 3 | 104 | 62 |
AY099469.1 | - | 1 - 2 | 217 | 62 | |
BC113119.1 | - | 1 - 2 | 202 | 62 | |
BC117701.1 | - | 1 - 2 | 202 | 62 | |
BP358360.1 | - | 1 - 2 | 231 | 62 |