Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD1C
Entrez Gene ID
911
Gene Name
CD1c molecule
Gene Aliases
BDCA1, CD1, CD1A, R7
Chromosome Location
Chr.1: 158289773 - 158294774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001765.2 NP_001756.2 4 - 5 1183 76
GenBank mRNA AK226024.1 - 4 - 5 1187 76
AK298234.1 - 4 - 5 1023 76
BC126465.1 - 4 - 5 922 76
BC126467.1 - 4 - 5 922 76
M28827.1 - 4 - 5 76

View Full Product Details