The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001895.2 | NP_001001895.1 | 10 - 11 | 1452 | 72 |
NM_001243467.1 | NP_001230396.1 | 10 - 11 | 1452 | 72 | |
NM_018961.3 | NP_061834.1 | 11 - 12 | 1566 | 72 | |
XM_006724013.3 | XP_006724076.1 | 11 - 12 | 1650 | 72 | |
XM_011529605.2 | XP_011527907.1 | 11 - 12 | 1566 | 72 | |
XM_011529606.2 | XP_011527908.1 | 11 - 12 | 1535 | 72 | |
XM_011529607.2 | XP_011527909.1 | 11 - 12 | 1619 | 72 | |
XM_011529610.2 | XP_011527912.1 | 7 - 8 | 3565 | 72 | |
GenBank mRNA | AF520809.1 | - | 11 - 12 | 1493 | 72 |
AF521702.1 | - | 10 - 11 | 1379 | 72 | |
AJ277750.1 | - | 11 - 12 | 1513 | 72 | |
AK308151.1 | - | 10 - 11 | 1392 | 73 | |
AK313367.1 | - | 10 - 11 | 1433 | 72 | |
BC069357.1 | - | 11 - 12 | 1513 | 72 | |
BC069483.1 | - | 11 - 12 | 1513 | 72 | |
BC069511.1 | - | 10 - 11 | 1397 | 72 | |
BC069577.1 | - | 10 - 11 | 1399 | 72 |