The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175058.4 | NP_778228.3 | 18 - 19 | 2532 | 90 |
XM_017017240.1 | XP_016872729.1 | 20 - 21 | 2740 | 90 | |
XM_017017241.1 | XP_016872730.1 | 20 - 21 | 2740 | 90 | |
XM_017017242.1 | XP_016872731.1 | 19 - 20 | 2620 | 90 | |
XM_017017243.1 | XP_016872732.1 | 18 - 19 | 2539 | 90 | |
GenBank mRNA | AK296371.1 | - | 8 - 9 | 1267 | 90 |
AK301465.1 | - | 8 - 9 | 1123 | 90 | |
BC033239.1 | - | 9 - 10 | 1409 | 90 | |
BC071599.1 | - | 18 - 19 | 2528 | 90 |