The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144063.1 | NP_001137535.1 | 13 - 14 | 1575 | 70 |
NM_020896.3 | NP_065947.1 | 14 - 15 | 1779 | 70 | |
NM_145638.2 | NP_663613.1 | 13 - 14 | 1568 | 70 | |
XM_011519873.2 | XP_011518175.1 | 14 - 15 | 2118 | 70 | |
XM_017017163.1 | XP_016872652.1 | 14 - 15 | 2126 | 70 | |
XM_017017164.1 | XP_016872653.1 | 9 - 10 | 1697 | 70 | |
GenBank mRNA | AB074006.1 | - | 14 - 15 | 1720 | 70 |
AF392453.1 | - | 14 - 15 | 1692 | 70 | |
AF410430.1 | - | 13 - 14 | 1637 | 70 | |
AK055617.1 | - | 9 - 10 | 1113 | 70 | |
AK056510.1 | - | 14 - 15 | 1737 | 70 | |
AK293000.1 | - | 13 - 14 | 1524 | 70 | |
AK297475.1 | - | 10 - 11 | 874 | 70 | |
AK301003.1 | - | 12 - 13 | 1415 | 70 | |
AK303205.1 | - | 14 - 15 | 1597 | 70 | |
AK309411.1 | - | 14 - 15 | 1757 | 70 | |
AL136918.1 | - | 14 - 15 | 1692 | 70 | |
BC017505.1 | - | 5 - 6 | 435 | 70 | |
BC032646.1 | - | 13 - 14 | 1489 | 70 | |
BC039579.1 | - | 14 - 15 | 1690 | 70 |