The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169117.1 | NP_001162588.1 | 1 - 2 | 676 | 57 |
NM_001169118.1 | NP_001162589.1 | 1 - 2 | 676 | 57 | |
NM_020860.3 | NP_065911.3 | 1 - 2 | 676 | 57 | |
GenBank mRNA | AB040915.1 | - | 1 - 2 | 364 | 57 |
AF328905.1 | - | 1 - 2 | 678 | 57 | |
AK096846.1 | - | 1 - 2 | 527 | 57 | |
AL136577.1 | - | 1 - 2 | 219 | 57 | |
BC136449.1 | - | 1 - 2 | 475 | 57 | |
BC146661.1 | - | 1 - 2 | 572 | 57 | |
BC152554.1 | - | 1 - 2 | 572 | 57 | |
BC171766.1 | - | 1 - 2 | 474 | 57 |