The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005822.3 | NP_005813.2 | 1 - 2 | 497 | 75 |
GenBank mRNA | AK312392.1 | - | 1 - 2 | 204 | 75 |
BC038509.1 | - | 1 - 2 | 280 | 75 | |
DA106611.1 | - | 1 - 2 | 497 | 75 | |
DA207634.1 | - | 1 - 2 | 486 | 75 |