The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012308.2 | NP_036440.1 | 7 - 8 | 1450 | 78 |
NR_027473.1 | 7 - 8 | 1450 | 78 | ||
XM_006718480.3 | XP_006718543.1 | 3 - 4 | 468 | 78 | |
XM_011544860.2 | XP_011543162.1 | 7 - 8 | 1509 | 78 | |
XM_011544861.1 | XP_011543163.1 | 2 - 3 | 187 | 78 | |
XM_017017391.1 | XP_016872880.1 | 7 - 8 | 756 | 78 | |
XM_017017392.1 | XP_016872881.1 | 6 - 7 | 567 | 78 | |
GenBank mRNA | AB023221.2 | - | 7 - 8 | 767 | 78 |
AK024505.1 | - | 7 - 8 | 692 | 78 | |
BC047371.1 | - | 2 - 3 | 187 | 78 | |
BC047486.1 | - | 7 - 8 | 1452 | 78 | |
BC064360.1 | - | 7 - 8 | 865 | 78 | |
JQ710741.1 | - | 6 - 7 | 634 | 78 | |
JQ710742.1 | - | 7 - 8 | 756 | 78 | |
JQ710743.1 | - | 6 - 7 | 634 | 78 | |
JQ710744.1 | - | 8 - 9 | 828 | 78 |