The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267039.1 | NP_001253968.1 | 14 - 15 | 2155 | 81 |
NM_015454.2 | NP_056269.1 | 12 - 13 | 1819 | 81 | |
NM_016648.3 | NP_057732.2 | 12 - 13 | 1947 | 81 | |
NR_049768.1 | 12 - 13 | 1844 | 81 | ||
GenBank mRNA | AF068284.1 | - | 11 - 12 | 1096 | 81 |
AK225900.1 | - | 9 - 10 | 1316 | 81 | |
BC107709.1 | - | 6 - 7 | 788 | 81 | |
BX647100.1 | - | 12 - 13 | 1681 | 81 | |
CA314598.1 | - | 1 - 2 | 81 |