The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135208.1 | NP_001128680.1 | 3 - 4 | 390 | 73 |
NM_004470.3 | NP_004461.2 | 3 - 4 | 475 | 73 | |
NM_057092.2 | NP_476433.1 | 3 - 4 | 381 | 73 | |
XM_005273848.3 | XP_005273905.1 | 3 - 4 | 377 | 73 | |
GenBank mRNA | AW450265.1 | - | 3 - 4 | 73 | |
BC003384.1 | - | 3 - 4 | 381 | 73 | |
BC091475.1 | - | 3 - 4 | 327 | 73 | |
BT009861.1 | - | 2 - 3 | 279 | 73 | |
BU682774.1 | - | 3 - 4 | 73 | ||
CR407636.1 | - | 2 - 3 | 279 | 73 | |
M65128.1 | - | 3 - 4 | 73 | ||
M75099.1 | - | 3 - 4 | 309 | 73 |