The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126328.2 | NP_001119800.1 | 5 - 6 | 1263 | 65 |
NM_032622.2 | NP_116011.2 | 4 - 5 | 994 | 65 | |
XM_005265785.4 | XP_005265842.1 | 5 - 6 | 1421 | 65 | |
XM_017008776.1 | XP_016864265.1 | 6 - 7 | 1482 | 65 | |
GenBank mRNA | AF237782.1 | - | 4 - 5 | 925 | 65 |
AK294611.1 | - | 5 - 6 | 792 | 65 | |
AY358326.1 | - | 4 - 5 | 905 | 65 | |
BC022983.1 | - | 5 - 6 | 1139 | 65 | |
BC034737.1 | - | 4 - 5 | 914 | 65 |