The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167604.1 | NP_001161076.1 | 10 - 11 | 1162 | 90 |
NM_001324128.1 | NP_001311057.1 | 8 - 9 | 1009 | 90 | |
NM_001324131.1 | NP_001311060.1 | 11 - 12 | 1257 | 90 | |
NM_001324132.1 | NP_001311061.1 | 9 - 10 | 1073 | 90 | |
NM_001324133.1 | NP_001311062.1 | 10 - 11 | 1162 | 90 | |
NM_001324134.1 | NP_001311063.1 | 8 - 9 | 993 | 90 | |
NM_001324135.1 | NP_001311064.1 | 10 - 11 | 1162 | 90 | |
NM_001324136.1 | NP_001311065.1 | 10 - 11 | 1377 | 90 | |
NM_020383.3 | NP_065116.3 | 10 - 11 | 1162 | 90 | |
XM_011540134.1 | XP_011538436.1 | 10 - 11 | 1156 | 90 | |
XM_017016613.1 | XP_016872102.1 | 9 - 10 | 1012 | 90 | |
XM_017016614.1 | XP_016872103.1 | 11 - 12 | 1145 | 90 | |
XM_017016615.1 | XP_016872104.1 | 9 - 10 | 1029 | 90 | |
GenBank mRNA | AF195530.1 | - | 9 - 10 | 989 | 90 |
AF272981.1 | - | 8 - 9 | 913 | 90 | |
AK093693.1 | - | 8 - 9 | 967 | 90 | |
AK223513.1 | - | 9 - 10 | 973 | 90 | |
AK289436.1 | - | 10 - 11 | 1065 | 90 | |
AK295756.1 | - | 2 - 3 | 418 | 90 | |
AK304367.1 | - | 10 - 11 | 1055 | 90 | |
AL833411.1 | - | 10 - 11 | 1068 | 90 | |
BC005126.2 | - | 9 - 10 | 951 | 90 | |
BC007579.1 | - | 10 - 11 | 1053 | 90 | |
BC013417.2 | - | 6 - 7 | 663 | 90 | |
BX537682.1 | - | 7 - 8 | 818 | 90 | |
CR456922.1 | - | 8 - 9 | 913 | 90 | |
X95762.1 | - | 8 - 9 | 913 | 90 |