Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYGL
Entrez Gene ID
5836
Gene Name
phosphorylase, glycogen, liver
Gene Aliases
GSD6
Chromosome Location
Chr.14: 50905217 - 50944530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163940.1 NP_001157412.1 3 - 4 548 99
NM_002863.4 NP_002854.3 4 - 5 650 99
GenBank mRNA AF046785.1 - 4 - 5 523 99
AF066858.1 - 4 - 5 523 99
AK225904.1 - 4 - 5 603 99
AK300580.1 - 3 - 4 501 99
AK309333.1 - 4 - 5 576 99
AK313206.1 - 4 - 5 564 99
BC009895.2 - 4 - 5 593 99
BC065011.1 - 4 - 5 590 99
BC082229.1 - 4 - 5 594 99
BC095850.1 - 4 - 5 610 99
BC110791.1 - 4 - 5 592 99
M14636.1 - 4 - 5 636 99
Y15233.1 - 4 - 5 574 99

View Full Product Details