The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178116.1 | NP_001171587.1 | 8 - 9 | 1091 | 70 |
NM_005470.3 | NP_005461.2 | 7 - 8 | 1040 | 70 | |
XM_005252332.3 | XP_005252389.1 | 6 - 7 | 840 | 70 | |
XM_006717359.2 | XP_006717422.1 | 7 - 8 | 939 | 70 | |
XM_006717360.2 | XP_006717423.1 | 7 - 8 | 939 | 70 | |
XM_011519289.2 | XP_011517591.1 | 7 - 8 | 939 | 70 | |
XM_017015458.1 | XP_016870947.1 | 7 - 8 | 939 | 70 | |
GenBank mRNA | AK126803.1 | - | 8 - 9 | 990 | 70 |
U87166.1 | - | 7 - 8 | 897 | 70 |