The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019043.3 | NP_061916.3 | 3 - 4 | 388 | 99 |
XM_006717451.2 | XP_006717514.1 | 2 - 3 | 86 | 99 | |
XM_011519514.2 | XP_011517816.1 | 3 - 4 | 401 | 99 | |
GenBank mRNA | AB085852.1 | - | 1 - 2 | 67 | 99 |
AK000812.1 | - | 3 - 4 | 347 | 99 | |
AK303597.1 | - | 3 - 4 | 948 | 99 | |
AK308916.1 | - | 3 - 4 | 369 | 99 | |
AY152730.1 | - | 2 - 3 | 154 | 99 | |
BC035636.1 | - | 3 - 4 | 300 | 99 | |
BC054516.1 | - | 2 - 3 | 166 | 99 | |
BM150270.1 | - | 3 - 4 | 389 | 99 | |
BM556115.1 | - | - | 355 | 99 |