Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC83
Entrez Gene ID
220047
Gene Name
coiled-coil domain containing 83
Gene Aliases
CT148, HSD9
Chromosome Location
Chr.11: 85855066 - 85920021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286159.1 NP_001273088.1 4 - 5 861 136
NM_173556.4 NP_775827.2 4 - 5 861 136
XM_011544839.2 XP_011543141.1 4 - 5 447 136
XM_011544840.2 XP_011543142.1 4 - 5 519 136
XM_011544841.1 XP_011543143.1 4 - 5 409 136
XM_011544842.1 XP_011543144.1 4 - 5 861 136
XM_011544844.2 XP_011543146.1 2 - 3 573 136
XM_017017354.1 XP_016872843.1 4 - 5 861 136
GenBank mRNA AK093405.1 - 2 - 3 241 136
AK124113.1 - 4 - 5 862 136
AK314036.1 - 4 - 5 565 136
BC040280.1 - 4 - 5 539 136

View Full Product Details