The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006397.2 | NP_006388.2 | 3 - 4 | 385 | 77 |
XM_006722619.2 | XP_006722682.1 | 4 - 5 | 570 | 77 | |
GenBank mRNA | AK315327.1 | - | 3 - 4 | 415 | 77 |
BC011748.2 | - | 3 - 4 | 376 | 77 | |
BG825340.1 | - | 3 - 4 | 389 | 77 | |
CR456841.1 | - | 3 - 4 | 325 | 77 | |
Z97029.1 | - | 3 - 4 | 385 | 77 |