The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013284.3 | NP_037416.1 | 8 - 9 | 1170 | 65 |
NR_104299.1 | 8 - 9 | 1141 | 65 | ||
XM_005249711.2 | XP_005249768.1 | 5 - 6 | 721 | 65 | |
XM_006715692.2 | XP_006715755.1 | 8 - 9 | 1141 | 65 | |
XM_006715696.2 | XP_006715759.1 | 6 - 7 | 871 | 65 | |
XM_006715698.3 | XP_006715761.1 | 3 - 4 | 1025 | 65 | |
XM_011515275.1 | XP_011513577.1 | 8 - 9 | 1156 | 65 | |
XM_011515278.1 | XP_011513580.1 | 7 - 8 | 1006 | 65 | |
XM_011515279.1 | XP_011513581.1 | 8 - 9 | 1156 | 65 | |
XM_011515282.2 | XP_011513584.1 | 3 - 4 | 1022 | 65 | |
XM_011515285.1 | XP_011513587.1 | 4 - 5 | 408 | 65 | |
XM_017011998.1 | XP_016867487.1 | 7 - 8 | 970 | 65 | |
XM_017012000.1 | XP_016867489.1 | 7 - 8 | 1006 | 65 | |
XM_017012002.1 | XP_016867491.1 | 7 - 8 | 986 | 65 | |
XM_017012003.1 | XP_016867492.1 | 5 - 6 | 734 | 65 | |
GenBank mRNA | AF176097.1 | - | 8 - 9 | 1110 | 65 |
AJ131891.2 | - | 8 - 9 | 1113 | 65 | |
AK023002.1 | - | 8 - 9 | 1116 | 65 | |
AK092903.1 | - | 4 - 5 | 893 | 65 | |
AK307634.1 | - | 5 - 6 | 1007 | 65 |