The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032782.4 | NP_116171.3 | 4 - 5 | 779 | 93 |
GenBank mRNA | AB924452.1 | - | 3 - 4 | 462 | 93 |
AF251707.1 | - | 4 - 5 | 651 | 93 | |
AF450242.1 | - | 4 - 5 | 582 | 93 | |
AF450243.1 | - | 4 - 5 | 582 | 93 | |
AK027334.1 | - | 4 - 5 | 568 | 93 | |
AK130144.1 | - | 2 - 3 | 76 | 93 | |
AK314406.1 | - | 4 - 5 | 586 | 93 | |
AY069944.1 | - | 4 - 5 | 525 | 93 | |
BC063431.1 | - | 4 - 5 | 669 | 93 | |
JX049979.1 | - | 4 - 5 | 525 | 93 |