The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201538.1 | NP_001188467.1 | 4 - 5 | 419 | 84 |
NM_001201539.1 | NP_001188468.1 | 4 - 5 | 504 | 84 | |
NM_004042.4 | NP_004033.2 | 4 - 5 | 385 | 84 | |
XM_011545522.1 | XP_011543824.1 | 4 - 5 | 371 | 84 | |
XM_011545523.1 | XP_011543825.1 | 4 - 5 | 371 | 84 | |
XM_011545524.1 | XP_011543826.1 | 4 - 5 | 371 | 84 | |
XM_017029527.1 | XP_016885016.1 | 4 - 5 | 385 | 84 | |
XM_017029528.1 | XP_016885017.1 | 4 - 5 | 371 | 84 | |
GenBank mRNA | BC022389.1 | - | 4 - 5 | 504 | 84 |
BP274206.1 | - | 4 - 5 | 419 | 84 | |
BP277542.1 | - | 4 - 5 | 417 | 84 | |
DA156994.1 | - | 4 - 5 | 386 | 84 | |
DA628671.1 | - | 4 - 5 | 418 | 84 | |
X97868.1 | - | 5 - 6 | 356 | 84 |