The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057175.3 | NP_476516.1 | 13 - 14 | 1788 | 110 |
XM_005263236.2 | XP_005263293.1 | 13 - 14 | 1843 | 110 | |
GenBank mRNA | AF327722.1 | - | 13 - 14 | 1811 | 110 |
AJ314788.1 | - | 13 - 14 | 1740 | 110 | |
AK304417.1 | - | 12 - 13 | 1469 | 110 | |
AK314626.1 | - | 13 - 14 | 1843 | 110 | |
AY039242.1 | - | 13 - 14 | 1813 | 110 | |
AY112670.1 | - | 13 - 14 | 1532 | 110 | |
BC032642.1 | - | 13 - 14 | 1813 | 110 | |
BC039818.1 | - | 13 - 14 | 1803 | 110 | |
BC093928.1 | - | 13 - 14 | 1597 | 110 | |
BC104806.1 | - | 13 - 14 | 1597 | 110 |