Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- KDM6A
- Entrez Gene ID
-
7403
- Gene Name
- lysine demethylase 6A
- Gene Aliases
-
KABUK2, UTX, bA386N14.2
- Chromosome Location
-
Chr.X: 44873175 - 45112612 on Build GRCh38
View Full Product Details