The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308339.1 | NP_001295268.1 | 5 - 6 | 1164 | 112 |
NM_144973.3 | NP_659410.3 | 3 - 4 | 1094 | 112 | |
XM_005253326.2 | XP_005253383.1 | 4 - 5 | 1096 | 112 | |
XM_005253327.3 | XP_005253384.1 | 3 - 4 | 1089 | 112 | |
XM_006719040.3 | XP_006719103.1 | 4 - 5 | 1096 | 112 | |
XM_017018896.1 | XP_016874385.1 | 4 - 5 | 1096 | 112 | |
XM_017018897.1 | XP_016874386.1 | 4 - 5 | 1459 | 112 | |
XM_017018898.1 | XP_016874387.1 | 4 - 5 | 1096 | 112 | |
GenBank mRNA | AK123642.1 | - | 1 - 2 | 511 | 112 |
AK125323.1 | - | 5 - 6 | 1164 | 112 | |
BC020855.1 | - | 2 - 3 | 764 | 112 | |
BC068580.1 | - | 4 - 5 | 1151 | 112 |